Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ dNTP Mix (2.5 mM)

Catalog No. r72501
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R72501 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R72501 Supplier Invitrogen™ Supplier No. R72501
Only null left
Add to Cart
Add to Cart

Includes

1mL (2 x 500μL) contains sufficient 2.5mM dNTPs for 200 PCR reactions (50μL each).

Invitrogen 2.5 mM dNTP (2'-deoxynucleoside 5'-triphosphate) Mix consists of a solution of all four nucleotides, dATP, dCTP, dGTP, and dTTP, each at a concentration of 2.5 mM.

Invitrogen 2.5 mM dNTP (2'-deoxynucleoside 5'-triphosphate) Mix consists of a solution of all four nucleotides, dATP, dCTP, dGTP, and dTTP, each at a concentration of 2.5 mM. It is supplied in purified water and neutralized to pH 8.0 with NaOH. 2.5 mM dNTP Mix is suitable for use in a wide variety of molecular applications.

Applications
• Traditional PCR
• Sequencing
• cDNA synthesis
• Nick translation
• TdT-tailing reactions

TRUSTED_SUSTAINABILITY

Specifications

Concentration 2.5 mM
Content And Storage dNTP Mix, 2.5 mM (2 x 500 μL)

Store at –20°C.
Form Liquid
Label or Dye Unlabeled
Quantity 1 mL
Shipping Condition Dry Ice
For Use With (Application) Standard PCR

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.