Learn More
Scientific Instrument Services Inc Inland 19 Pump Oil, 1 Gallon Bottle
Supplier: Scientific Instrument Services Inc IN191
Inland rough pump oil is ideal for use in all models of vacuum pumps including Adixen, Edwards, and Welch. Inland 19 utilizes crude stocks known for their durability and fine lubricating qualities. Precision distillation further enhances these characteristics while providing a cleaner, cooler running, longer-lasting oil that in many instances will provide improved pumping cycles. Inland 19 is effective in those cases where operating temperatures are not a problem, especially in the lower speed belt-driven pumps. Providing the least back streaming, Inland 19 is particularly well suited for use with the high grade diffusion pump fluids, such as the polyphenyl ethers (Santovac 5 and Convalex 10), silicones, and ester-based fluids. Direct-drive pumps that are not subject to extreme operating conditions can use Inland 19 to achieve the same advantages. Durability and performance highlight Inland 19's capabilities. Inland 19 replaces Inland 15.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.