Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Imatinib 98.0+%, TCI America™
SDP

Catalog No. I0906100MG Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
100 mg
1 g

Catalog No. I0906100MG

Supplier: TCI America I0906100MG

Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 152459-95-5
Molecular Formula C29H31N7O
Molecular Weight (g/mol) 493.62
MDL Number MFCD05662257
InChI Key KTUFNOKKBVMGRW-UHFFFAOYSA-N
PubChem CID 5291
ChEBI CHEBI:45783
IUPAC Name N-(4-methyl-3-{[4-(pyridin-3-yl)pyrimidin-2-yl]amino}phenyl)-4-[(4-methylpiperazin-1-yl)methyl]benzamide
SMILES CN1CCN(CC2=CC=C(C=C2)C(=O)NC2=CC=C(C)C(NC3=NC=CC(=N3)C3=CC=CN=C3)=C2)CC1

Specifications

Melting Point 209°C
Quantity 100 mg
Formula Weight 493.62
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Imatinib
RTECSNumber CV5585673
TSCA No
Recommended Storage Freezer
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.