Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL17RE Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589747
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA589747 100 μL
1 options

Catalog No. PIPA589747

Supplier: Invitrogen™ PA589747

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AGIGFRHLPH WNTRCPLASH TDDSFTGSSA YIPCRTWWAL FSTKPWCVRV WHCSRCLCQH LLSGGSGLQR GLFHLLVQKS KKSSTFKFYR RHKMPAPAQR KLLPRRHLSE KSHHISIPSP DISHKGLRSK RTQPSDPETW ESLPRLDSQR HGGPEFSFDL LPEARAIRVT ISSGPEVSVR LCHQWALECE ELSSPYDVQK IVSGGHTVEL PYEFLLPCLC IEASYLQEDT VRRKKCP; Positive Samples: HT-29, A-549, A-431, Mouse kidney, Mouse thymus; Cellular Location: Cell membrane, Cytoplasm, Single-pass type I membrane protein.

The interleukin 17 receptor E (IL-17RE) shares limited similarity to the receptor of IL-17A, a ubiquitous type I membrane glycoprotein that with IL-17A plays a pathogenic role in many inflammatory and autoimmune diseases such as rheumatoid arthritis. IL-17RE is the functional receptor of IL-17C and has been suggested to mediate mucosal immunity to infection with intestinal pathogens. IL-17C is produced by epithelia in response to bacterial challenge and binds to a receptor complex formed by IL-17RE and IL-17RA on the epithelial cell surface, thereby regulating the immune function of epithelial cells in an autocrine manner.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL17RE
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.98 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene IL17RE
Gene Accession No. Q8BH06, Q8NFR9
Gene Alias AA589509; IL-17 receptor E; IL17RE; IL-17RE; Il25r; interleukin 17 receptor E; interleukin-17 receptor E; UNQ3056/PRO9877
Gene Symbols IL17RE
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 24-260 of human IL17RE (NP_7056131).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 132014, 57890
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.