Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ eBioscience™ IHC /ICC Blocking Buffer - Low Protein

Catalog No. 5018486
Encompass_Preferred
Click to view available options
Quantity:
50 mL

Catalog No. 50-184-86

Supplier: Invitrogen™ 00495354

Only null left
Add to Cart
Add to Cart

Designed for use in immunohistochemistry and immunocytochemistry protocols which require blocking of non-specific binding sites. IHC /ICC Blocking Buffer, Low Protein is recommended for tissues and antibodies with average levels of background staining.

  • Can be used both in the blocking step and as a diluent for unconjugated antibodies and organic dye-conjugated antibodies during the antibody incubation step
  • Supplied as a 1X stock solution containing sodium azide
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Blocking Buffer
For Use With (Application) Immunocytochemistry (ICC)
Physical Form Liquid
Product Line eBioscience
Quantity 50 mL

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.