Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZeptoMetrix Acinetobacter lwoffii Z141, Titered
Shop All ZeptoMetrix Corporation Products

Click to view available options
Quantity:
1 mL
Description
- Identification of organism was confirmed by 16S sequencing
- Purity of the culture was monitored by Gram staining and by additional culturing
- Freezing medium contains 15% glycerol as a cryoprotectant
Specifications
Specifications
| Biosafety Level | 2 |
| Certifications/Compliance | ISO 13485 |
| Content And Storage | -65°C or below |
| Form | Solution |
| Product Type | Titered Live Bacteria Control |
| For Use With (Application) | Determine LOD in diagnostic assay development/cross-reactivity studies |
| Quantity | 1 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction