Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hygiena SuperSnap ATP Test
SDP

Catalog No. SUS3000
Click to view available options
Quantity:
100/Pk.

Catalog No. SUS3000

Supplier: Hygiena SUS3000

Only null left
Add to Cart
Add to Cart

SuperSnap is capable of detecting extremely low levels of ATP, making it ideal for supporting stringent SOPs, including allergen cross-contamination prevention.

  • Easy to use - Just swab, snap, and squeeze
  • Get results in seconds, take immediate corrective actions and get back to production
  • Compatible with all Hygiena luminometers
  • Superior Sample Collection
  • Patented Snap-Valve Design
  • Liquid-Stable Chemistry

Specifications

Product Type ATP Surface Test
For Use With (Application) Used predominantly in food and beverage processing facilities
Quantity 100/Pk.
Storage Requirements 2°C to 8°C, 21°C to 25°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.