Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hydrogen peroxide, 27% w/w aq. soln., stab.
Click to view available options
Quantity:
2 x 50 mL
500 mL
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Specifications
Specifications
| Boiling Point | 107°C |
| Melting Point | -26°C |
| UN Number | UN2014 |
| Concentration | 27% w/w aq. Soln |
| Beilstein | 3587191 |
| Density | 1.1 |
| Merck Index | 14,4798 |
| Solubility Information | Miscible with water,ether,alcohol,organic solvents and carboxylic esters. Immiscible with hydrocarbons and petroleum ether. |
| Formula Weight | 34.02 |
| Refractive Index | 1.335 |
| Show More |
Safety and Handling
| Hazard Category | H271-H302+H332-H314 |
| Hazard Statement | GHS H Statement H272-H318-H302 May intensify fire; oxidizer. Causes serious eye damage. Harmful if swallowed. |
| Precautionary Statement | P210-P220-P221-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: HYDROGEN PEROXIDE, AQUEOUS SOLUTIONS |
| EINECSNumber | 231-765-0 |
| RTECSNumber | MX0899000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction