Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human VSIG1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$164.60 - $402.34
Specifications
| Antigen | VSIG1 |
|---|---|
| Clone | 986709 |
| Dilution | Western Blot 2 ug/mL, Simple Western 20 ug/mL, Immunohistochemistry 0.5-25 ug/mL |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB48182SP
![]() |
R&D Systems
MAB48182SP |
25 μg |
Each for $164.60
|
|
|||||
MAB48182100
![]() |
R&D Systems
MAB48182100 |
100 μg |
Each for $402.34
|
|
|||||
Description
VSIG1 Monoclonal specifically detects VSIG1 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.Specifications
| VSIG1 | |
| Western Blot 2 ug/mL, Simple Western 20 ug/mL, Immunohistochemistry 0.5-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Cell surface A33 antigen, dJ889N15.1, Glycoprotein A34, GPA34, MGC44287,1700062D20Rik, V-set and immunoglobulin domain containing 1, V-set and immunoglobulin domain-containing protein 1 | |
| Mouse myeloma cell line NS0-derived recombinant human VSIG1, Val22-Gly234, Accession # Q86XK7 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 986709 | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q86XK7 | |
| 340547 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title