Learn More
Biolegend Human TruStain FcX™ (Fc Receptor Blocking Solution) 200 Tests
Supplier: Biolegend 422302
Human Fc receptors (FcRs) are expressed on a variety of cells, such as monocytes, granulocytes, B cells and dendritic cells. The cells with FcR expression sometimes give false positive or false negative results of immunofluorescent staining due to the FcRs-mediated Ig Fc binding. Human TruStain FcX™ is specially formulated for blocking the FcR-involved unwanted staining without interfering with antibody-mediated specific staining of human cells. Human TruStain FcX™ is compatible with flow cytometric staining with anti-human CD16 (clone 3G8), CD32 (clone FUN-2), and CD64 (clone 10.1) antibodies.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.