Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human SSC5D (aa 870-962) Control Fragment Recombinant Protein

Catalog No. RP99201
Encompass_Preferred
Click to view available options
Quantity:
100 μL

Catalog No. RP99201

Supplier: Invitrogen™ RP99201

Only null left
Add to Cart
Add to Cart

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (65%), Rat (65%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-59819 (PA5-59819. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Binds to extracellular matrix proteins. Binds to pathogen-associated molecular patterns (PAMPs) present on the cell walls of Gram-positive and Gram-negative bacteria and fungi, behaving as a pattern recognition receptor (PRR). Induces bacterial and fungal aggregation and subsequent inhibition of PAMP-induced cytokine release. Does not possess intrinsic bactericidal activity. May play a role in the innate defense and homeostasis of certain epithelial surfaces.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number A1L4H1
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 284297
Name Human SSC5D (aa 870-962) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias S5D-SRCRB; scavenger receptor cysteine rich domain containing (5 domains); scavenger receptor cysteine rich family member with 5 domains; scavenger receptor cysteine rich family, 5 domains; scavenger receptor cysteine-rich glycoprotein; soluble scavenger protein with 5 SRCR domains; Soluble scavenger receptor cysteine-rich domain-containing protein SSC5D; soluble scavenger with 5 domains; SSC5D
Common Name SSC5D
Gene Symbol SSC5D
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence TCTGYTDYDDYPPWTWDPTSREDLAKGTTTAGVPGHTLPWRTTRRPGSSSPAIRRLPDTGSKDGYKLPWTWDTPSGRGLAEGTPTAGKLGPTL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.