Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Human Pro-MMP-1 Quantikine ELISA Kit
Click to view available options
Quantity:
1 Pk. (6 Plates)
1 Plate
Sufficient For:
1 x 96-well Plate
6 x 96-well Plates
Description
The Human Pro-MMP-1 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human MMP-1 in cell culture supernates (100 ul), serum (100 ul), heparin plasma (100 ul).
Specifications
Specifications
| Assay Range | 0.16 to 10 ng/mL |
| Assay Sensitivity | 0.095 ng/mL |
| Conjugate | HRP |
| Cross Reactivity | Cross-species reactivity not tested. <0.5% cross-reactivity observed with available related molecules. |
| Product Type | Solid Phase Sandwich ELISA |
| Sample Type | Cell Culture Supernatant, Plasma (Heparin), Serum |
| Specificity | Natural and Recombinant Human Pro-MMP-1 |
| Sufficient For | 6 x 96-well Plates |
| Target Species | Human |
| For Use With (Application) | ELISA |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction