Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human PMFBP1 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$164.60 - $437.75
Specifications
| Antigen | PMFBP1 |
|---|---|
| Clone | 2310A |
| Dilution | Immunohistochemistry 3-25 μg/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB9967SP
![]() |
R&D Systems
MAB9967SP |
25 μg |
Each for $164.60
|
|
|||||
MAB9967100
![]() |
R&D Systems
MAB9967100 |
100 μg |
Each for $437.75
|
|
|||||
Description
PMFBP1 Monoclonal specifically detects PMFBP1 in Human samples. It is validated for Immunohistochemistry.Specifications
| PMFBP1 | |
| Immunohistochemistry 3-25 μg/mL | |
| Monoclonal | |
| Supernatant | |
| Human | |
| Q8TBY8 | |
| 83449 | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 2310A | |
| Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| DKFZp434G131, F77, FLJ40146, ODF3, PMF-1 binding protein, PMF-1-binding protein, polyamine modulated factor 1 binding protein 1, polyamine-modulated factor 1-binding protein 1, Stap | |
| E. coli-derived recombinant human PMFBP1 Asp737-Lys839 Accession # Q8TBY8 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human PMFBP1 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title