Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-2 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF202SP
This item is not returnable.
View return policy
Description
IL-2 Polyclonal specifically detects IL-2 in Human samples. It is validated for Western Blot, ELISA, Neutralization, Immunocytochemistry.Specifications
| IL-2 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| aldesleukin, IL2, IL-2, IL-2lymphokine, interleukin 2, interleukin-2, involved in regulation of T-cell clonal expansion, Proleukin, T cell growth factor, T-cell growth factor, TCGF | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3558 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Neutralization, Immunocytochemistry | |
| LYOPH | |
| Western Blot 0.5 ug/mL, Neutralization ≤0.15 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| P60568 | |
| IL2 | |
| E. coli-derived recombinant human IL-2 Ala21-Thr153 Accession # P60568 | |
| 25 μg | |
| Primary | |
| Detects human IL-2 in direct ELISAs and Western blots. In direct ELISAs, greater than 50% cross-reactivity with recombinant rabbit IL-2 is observed and less than 20% cross-reactivity with recombinant bovine IL-2, recombinant mouse IL-2 and recombinant rat IL-2 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction