Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-10 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$526.00
Specifications
Antigen | IL-10 |
---|---|
Clone | 2050B |
Dilution | Neutralization 75-450 ng/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells |
Applications | Neutralization, Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB9210100
![]() |
R&D Systems
MAB9210100 |
100 μg |
Each for $526.00
|
|
|||||
MAB9210SP
|
R&D Systems
MAB9210SP |
25 μg | N/A | N/A | N/A | ||||
Description
IL-10 Monoclonal specifically detects IL-10 in Human samples. It is validated for Neutralization, Intracellular Staining by Flow Cytometry.Specifications
IL-10 | |
Neutralization 75-450 ng/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
CSIF, CSIFMGC126450, Cytokine synthesis inhibitory factor, GVHDS, IL10, IL10A, IL-10MGC126451, interleukin 10, interleukin-10, TGIF | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-10 Met1-Asn178 Accession # P22301 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human IL-10 in direct ELISAs. |
2050B | |
Neutralization, Flow Cytometry | |
Unconjugated | |
Rabbit | |
Human | |
P22301 | |
3586 | |
IgG2b | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
RUO
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title