Learn More
Innovative Research Inc Human IgG ELISA Kit
Supplier:  Innovative Research Inc IHUIGGKT
Encompass_Preferred
This human immunoglobulin G antigen assay is intended for the quantitative determination of total human IgG antigen in serum, plasma, hybridoma cell supernatants, ascites or other biological fluids. The assay does not distinguish IgG subclasses. For research use only. Strip well format. Reagents for up to 96 tests.
Catalog No. 50-203-6117