Learn More
ACROBiosystems Human IFN-alpha 1 Protein, His Tag
Supplier: ACROBiosystems IFAH52H91MG
Recombinant Protein;1MG;Human IFN-alpha 1, His Tag (IFA-H52H9) is expressed from human 293 cells (HEK293). It contains AA Cys 24 - Glu 189 (Accession # P01562-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 21.3 kDa. The protein migrates as 22-24 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.