Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human GASP-1/WFIKKN2 Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $549.50
Specifications
| Antigen | GASP-1/WFIKKN2 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL, Neutralization 18.9 ug/mL |
| Applications | Western Blot, Immunoprecipitation, Neutralization |
| Classification | Polyclonal |
Description
GASP-1/WFIKKN2 Polyclonal specifically detects GASP-1/WFIKKN2 in Human samples. It is validated for Western Blot, Immunoprecipitation, Neutralization.Specifications
| GASP-1/WFIKKN2 | |
| Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL, Neutralization 18.9 ug/mL | |
| Polyclonal | |
| Goat | |
| Human | |
| GASP1, GASP-1, Growth and differentiation factor-associated serum protein 1, hGASP-1, WAP, follistatin, immunoglobulin, kunitz and NTR domain-containing-relatedprotein, WAP, follistatin/kazal, immunoglobulin, kunitz and netrin domain containing 2, WAP, FS, Ig, two KU and NTR module related protein, WAP, kazal, immunoglobulin, kunitz and NTR domain-containing protein 2, WFIKKN2, WFIKKN-related protein, WFIKKNRP, WFIKKNRPmultivalent protease inhibitor protein | |
| WFIKKN2 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot, Immunoprecipitation, Neutralization | |
| Unconjugated | |
| RUO | |
| Q8TEU8 | |
| 124857 | |
| Mouse myeloma cell line NS0-derived recombinant human GASP-1/WFIKKN2 Leu35-His576 Accession # Q8TEU8 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human GASP-1/WFIKKN2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant human GASP-2 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title