Learn More
FUJIFILM IRVINE SCIENTIFIC INC Human EGF, Recombinant Protein, Shenandoah Biotechnology, 500 ug vial
Supplier:  FUJIFILM IRVINE SCIENTIFIC INC 10026500UG
Encompass_Preferred
Epidermal growth factor (EGF) is a growth factor that stimulates the proliferation, differentiation, and survival of epithelial and epidermal cells. EGF contains three intramolecular disulfide bonds and binds in high affinity to the epidermal growth factor receptor (EGFR). EGF is overexpressed in multiple tumor cell lines and promotes resistance to chemotherapy and radiation treatments.
Catalog No. 50-990-732
Only null left
Edge
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.