Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Human Cystatin C Quantikine ELISA Kit
Click to view available options
Sufficient For:
1 x 96-well Plate
Description
The Human Cystatin C Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human Cystatin C in cell culture supernates (50 ul), serum (50 ul), edta plasma (50 ul), heparin plasma (50 ul), saliva (50 ul), urine (50 ul), human milk (50 ul).
Specifications
Specifications
| Assay Range | 3.12 - 100ng/mL |
| Assay Sensitivity | 0.227ng/mL |
| Conjugate | HRP |
| Sample Type | Cell Culture Supernatant, Human Milk, Plasma (EDTA, Heparin), Saliva, Serum, Urine |
| Specificity | Natural and Recombinant Human Cystatin C |
| Sufficient For | 1 x 96-well Plate |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 1471 |
| Sample Volume | Cell Culture Supernates (50uL),Serum (50uL),EDTA Plasma (50uL),Heparin Plasma (50uL),Saliva (50uL),Urine (50uL),Human Milk (50uL) |
| Storage Requirements | Store the unopened product at 2° to 8°C. Do not use past expiration date. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction