Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human BMP-4 MAb (Clone 66119), R&D Systems™
GREENER_CHOICE

Catalog No. MAB757100
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
Catalog No. Quantity
MAB757100 100 μg
MAB757SP 25 μg
MAB757500 500 μg
3 options
This item is not returnable. View return policy

Catalog No. MAB757100

Supplier: R&D Systems MAB757100

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 12 publications

BMP-4 Monoclonal specifically detects BMP-4 in Human samples. It is validated for Western Blot, Neutralization.

Specifications

Antigen BMP-4
Applications Western Blot, Neutralization
Classification Monoclonal
Clone 66119
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Neutralization 1-3 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P12644
Gene Alias BMP2B, BMP-2B, BMP2B1, BMP2BMCOPS6, BMP4, BMP-4, Bone morphogenetic protein 2B, bone morphogenetic protein 4, DVR4, MCOPS6, OFC11, ZYME
Gene Symbols BMP4
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human BMP-4 Ser293-Arg408 Accession # P12644
Purification Method Protein A or G purified from ascites
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 652
Test Specificity Detects human BMP-4 in direct ELISAs and Western blots. This antibody shows approximately 5% cross-reactivity with recombinant human BMP-2.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.