Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human ADAM28 Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

$188.28 - $449.33

Specifications

Antigen ADAM28
Clone 1004266
Dilution Flow Cytometry 0.25 μg/106 cells, CyTOF-ready
Applications Flow Cytometry, CyTOF
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB93252SP
GREENER_CHOICE
View Documents
R&D Systems
MAB93252SP
25 μg
Each for $188.28
Only null left
Add to Cart
 
MAB93252100
GREENER_CHOICE
View Documents
R&D Systems
MAB93252100
100 μg
Each for $449.33
Only null left
Add to Cart
 
Description

Description

ADAM28 Monoclonal specifically detects ADAM28 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications

Specifications

ADAM28
Flow Cytometry 0.25 μg/106 cells, CyTOF-ready
Monoclonal
Supernatant
Human
Q0UKQ2
10863
IgG1
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
1004266
Flow Cytometry, CyTOF
Unconjugated
Mouse
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
a disintegrin and metalloproteinase domain 28, ADAM metallopeptidase domain 28, ADAM23MDC-L, disintegrin and metalloproteinase domain-containing protein 28, Dtgn1, EC 3.4.24, EC 3.4.24.-, eMDC II, eMDCII, Epididymial metalloproteinase-like, disintegrin-like, and cysteine-rich proteinII, MDCL, MDCLADAM 28, MDC-Lm, MDC-Ls, Metalloproteinase-like, disintegrin-like, and cysteine-rich protein L, metalloproteinase-like, disintegrin-like, and cysteine-rich protein-L, Protein-L
Chinese Hamster Ovary cell line, CHO-derived human ADAM28, Leu72 & Leu191-Ala623,, Accession # Q0UKQ2
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human ADAM28 in direct ELISAs.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.