Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HSF1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 7 publications
$214.28 - $617.01
Specifications
| Antigen | HSF1 |
|---|---|
| Dilution | Western Blot 1:1000 (colorimetric), ELISA, Immunoprecipitation 1:10-1:500, Gel Super Shift Assays 1:1-1:100, Enzyme Immunoassay |
| Applications | ELISA, Western Blot, Gel Shift, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB018778
![]() |
Novus Biologicals
NBP1974750.025MG |
0.025 mg |
Each for $214.28
|
|
|||||
NBP19747505
![]() |
Novus Biologicals
NBP1974750.05MG |
0.05 mg |
Each for $370.21
|
|
|||||
NBP19747502
![]() |
Novus Biologicals
NBP1974750.2MG |
0.2 mg |
Each for $617.01
|
|
|||||
Description
HSF1 Polyclonal specifically detects HSF1 in Human, Mouse, Rat, Gerbil, Primate, Rabbit samples. It is validated for Western Blot, ELISA, Immunoprecipitation, Gel Super Shift Assays, Enzyme Immunoassay.Specifications
| HSF1 | |
| ELISA, Western Blot, Gel Shift, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Primate, Rat, Rabbit, Human, Mouse, Gerbil | |
| Q00613 | |
| 3297 | |
| Recombinant human HSF1. | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot 1:1000 (colorimetric), ELISA, Immunoprecipitation 1:10-1:500, Gel Super Shift Assays 1:1-1:100, Enzyme Immunoassay | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS and 50% Glycerol with 0.09% Sodium Azide | |
| heat shock factor protein 1, heat shock transcription factor 1HSTF1HSF 1, HSTF 1 | |
| HSF1 | |
| IgG | |
| Protein A purified | |
| Recognizes nonphosphorylated HSF1 at ∼60kDa and phosphorylated HSF1 at ∼80kDa. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title