Learn More
Grainger 5/16" Wide, Interlocked Worm Gear Hose Clamp, 10/pk
Supplier: Grainger 1A527
1/4 to 5/8 In SAE 4 SS. Interlocked worm gear hose clamp helps secure many types of hoses, including pneumatic and exhaust hoses. This general-purpose hose clamp is interlocked to prevent shearing, while clamps used for exhaust have no holes, preventing marring of the hose and fumes escaping. Gear clamp resists rust and corrosion and are useful in humid or damp conditions. 10/pk
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.