Learn More
Simport™ Scientific Histosette™ II Biopsy Processing/Embedding Cassettes in Quickload™ Stacks
Shop All Simport Scientific Products
Click to view available options
Color:
Aqua
Gray
Lilac
Orange
Peach
Pink
Tan
Description
- In Quickload™ sleeve
- 45° anterior writing area
- Open and close as often as necessary - cassettes will always relock securely
- Back mounted tab allowing an unobstructed anterior marking area
- Suitable for automated printers
- Made of acetal
- Resistant to the chemical action of most histological solvents
Order Info
Each case contains 50 Quickload™ stacks of 40 cassettes.
Certifications
CE-IVD
Specifications
Specifications
| Quantity | 2000/Pk. |
| Color | Pink |
| Material | Acetal |