Learn More
Bioassay Systems EnzyChrom™ Histamine Assay Kit. For quantitative determination of histamine in food, beverage and agricultural products.
Supplier: Bioassay Systems EHIS100
For quantitative determination of histamine in food, beverage and agricultural products. Key Features: Fast and sensitive. Use of 20 µL sample. Linear detection range 0.0025 - 1 mM (0.3 - 111 ppm) histamine in 96-well plate assay. Convenient. The procedure involves adding a single working reagent, and reading the absorbance after 30 minutes. Room temperature assay. No 37°C heater is needed. High-throughput. "Add-mix-read" type assay. Can be readily automated as a high-throughput 96-well plate assay for thousands of samples per day. Method: OD565nm. Samples: Food, beverage and agricultural products. Species: All. Procedure: Assay takes 30 min. Kit size: 100 tests. Detection limit: 2.5 µM.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.