Learn More
Cole-Parmer High-Level Chlorine Test Strips, 0 to 10, 000 ppm; 50/Pk
Supplier:  Cole-Parmer 1810501
Encompass_Preferred
Cole-Parmer High-Level Chlorine Test Strips, 0 to 10, 000 ppm; 50/Pk, Broad-range test strips handle high-level available chlorine testing up to 10,000 ppm, These strips are perfect for testing the available chlorine in bleach solutions—great for conformance to CDC surface disinfection recommendations. Check available chlorine at 0, 1000, 2500, 5000, 7500, and 10,000 ppm. Each test takes approximately 30 seconds. Packaged in a vial of 50 strips. Each vial is lined with desiccant to maintain freshness
Catalog No. NC1232956
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.