Learn More
Cole-Parmer CN-200D Digital Heating Controller, 1500 W; 230 VAC
Supplier: Cole-Parmer 3258700
Versatile benchtop digital heating controller can be used in three ways: in on/off mode with the hysteresis loop controlling power switching, as a PID controller, or simply as a temperature measuring device using a probe. Sample temperatures are displayed on an easy-to-read LED display. Included PTFE-covered thermometer probe is suitable for measurements up to 250C or order optional temperature probe 32587-98 for measurements up to 800C. There is a 5-pin DIN socket on the controller for the probe. Rugged zinc die-cast outer housing is suitable for benchtop use or wall mounting. Rear clamp accepts a standard 1/2" (12.5 mm) rod to mount on a support stand or frame. .
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.