Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Heat Shrink Tubing

Catalog No. 19072057 Shop All 3M Products
Click to view available options
Quantity:
5 Kits/Cs.

Catalog No. 19-072-057

Supplier: 3M™ 7000031588

Only null left
Add to Cart
Add to Cart

3M™ Heat Shrink Tubing is a general purpose, thin wall tubing that provides a balance of electrical, physical and chemical properties, making it ideal for industrial and military applications.

  • Balances electrical, physical and chemical properties for various industrial and military applications
  • Offers shrink-fit protection and strain relief of wires, solder joints, terminals and connections
  • 2:1 shrink ratio
  • Suitable for lightweight wire-harness covering, fire-resistant covering, wire marking, wire bundling and component packaging
  • Withstands temperatures of -67 to 275°F (-55 to 135°C)
  • Flame-retardant tubing is rated for 600V applications
  • Flexible polyolefin construction that offers moisture seal protection.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Heat Shrink Tubing
For Use With (Application) Automotive, Design and Construction, Energy, Manufacturing, Mining, Oil and Gas, Transportation
Color Multi-Color
Model FPASSTAS6″K
Quantity 5 Kits/Cs.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.