Learn More
Cole-Parmer Megger MJ159 Analog Insulation Tester, Hand-Crank
Supplier: Cole-Parmer 2002258
These insulation resistance testers are the recognized standard in the industry. They have proven so reliable and popular that they have been reintroduced, generation after generation, with circuitry upgraded to complement the familiar design.True analog movement remains a popular feature, especially where diagnosis is involved. The trained eye can get much more than just a M reading from an analog meter. Similarly, a guard terminal adds a valuable extra dimension when more than just pass/fail testing is desired. With commonly four test voltages, extra range to the resistance measurements, continuity and k functions for proofing and troubleshooting, they address the widest range of applications and are the most general models.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.