Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gymnodimine Solution, 20 μg/g in Methanol (nominal concentration), Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111017113 Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
0.5 mL

Catalog No. 11-101-7113

Supplier: MilliporeSigma Supelco 807790.5ML

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to the SI through a primary reference material from a NMI.Certified content incl. uncertainty and expiry date are stated on the enclosed certificate.

TRUSTED_SUSTAINABILITY

Specifications

CAS 173792-58-0
Product Line TraceCERT
Concentration 20 μg/g in methanol (nom inal concentration)
Linear Formula C32H45NO4
MDL Number MFCD08435951
Quantity 0.5 mL
Formula Weight 507.70
Grade TraceCERT™ Analytical Standard

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.