Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GSC Go Science Crazy Dynamics Cart
SCI_ED

Catalog No. S13616 Shop All GSC Products
Encompass
Click to view available options
Color:
Blue

Not a toy. Product intended for ages 13 and up.

Catalog No. S13616

Supplier: GSC Go Science Crazy 460501

Only null left
Add to Cart
Add to Cart

Not a toy. Product intended for ages 13 and up.

A great tool for experiments in conservation, velocity, elastic collision, acceleration and momentum. The GSC Dynamics Cart is for performing experiments in conservation, velocity, elastic collision, acceleration and momentum. The set contains two high impact plastic cars with snap in wheels and two spring steel bumpers with rubber stopper assemblies. Each car has a recession for holding additional weight.

  • 2 Cars
  • Used for a variety of experiments
  • Recession for holding weight

Specifications

Description Set of two cars
For Use With (Application) For performing experiments in conservation, velocity, elastic collision, acceleration and momentum
Includes Recession for holding weight
Dimensions (L x W) 127 x 50 mm
Color Blue
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.