Learn More
World Precision Instrument Grip Strength Meter
Supplier: World Precision Instrument II2200
Measure muscle hyperalgesia in rats and mice with the Grip Strength Meter, which gauges the forelimb grip force using a digital force transducer. Simply hold the animal by the tail and gently dangle it over the wire mesh plate until the animal grasps the plate with its forepaws. The force transducer, connected with the wire mesh plate, measures the strength of the animal at the time of the test. The battery-operated, electronic control device calculates the average of three measurements, and it holds the last maximum force in a “peak and hold” type readout until you reset it.