Learn More
GA International Cryo-DTermo™ – DYMO-Compatible Cryogenic Labels
Supplier: GA International EG1F081GA
DYMO-compatible direct thermal labels are ideal for short and long-term cryo storage in liquid or vapor phase liquid nitrogen (-196°C/-321°F), laboratory freezers (-80°C/-112°F), and dry ice transportation. These freezer labels are waterproof and can be immersed in liquid nitrogen immediately after applying. Furthermore, their permanent cryo-adhesive ensures they retain adhesion and can withstand multiple freeze-thaw cycles as well as sterilization by gamma irradiation (up to 50 KGy).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.