Learn More
GPR43 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5111780
Description
Reconstitute antibody with 50 μL of double distilled water (DDW). The reconstituted solution can be stored at 4 degrees C for up to 1 week. For longer periods, small aliquots should be stored at -20 degrees C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min). Peptide sequence is: (C)RGAEETV EGTKTDR Homology: Mouse -12/14 amino acid residues identical.
G protein-coupled receptors (GPCRs, or GPRs) contain 7 transmembrane domains and transduce extracellular signals through heterotrimeric G proteins. GPCR GPR43 is a member of this family (subfamily carboxylic acid). It expression has been reported primarily in immune cells including monocytes and neutrophils. An EST has been isolated from a human pancreatic library.
Specifications
| GPR43 | |
| Polyclonal | |
| Unconjugated | |
| FFAR2 | |
| FFA2; FFA2R; Ffar2; Free fatty acid receptor 2; GPCR43; Gpr43; G-protein coupled receptor 43; Leukocyte-specific STAT-induced GPCR; Lssig | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 233079, 292794 | |
| -20°C | |
| Lyophilized |
| Western Blot | |
| 0.8 mg/mL | |
| PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
| Q76EI6, Q8VCK6 | |
| FFAR2 | |
| Peptide corresponding to amino acid residues 301-314 of rat FFAR2 (Intracellular, C-terminus) | |
| 50 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.