Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Themis, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of Themis.
Supplier: Abnova Corporation PAB18647
Description
Sequence: C-EERTINLPKSLKSHSpecifications
| Themis | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:64000) Western Blot (1-3 ug/mL) The optimal working dilution should be determined by the end user. | |
| E430004N04Rik | |
| A430108D01/BB160712 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 210757 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of Themis. | |
| In 0.5mg/mL Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA) | |
| NP_848781.1 | |
| E430004N04Rik | |
| A synthetic peptide corresponding to amino acids at C-terminus of mouse Themis. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction