Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Foxi3, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of Foxi3.
Supplier: Abnova Corporation PAB19732
Description
Sequence: C-RLRVSGKLEGDSPSSpecifications
| Foxi3 | |
| Polyclonal | |
| Goat polyclonal antibody raised against synthetic peptide of Foxi3. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| NP_001094934.1 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 232077 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:8000) Western Blot (1-3 ug/mL) The optimal working dilution should be determined by the end user. | |
| Foxi3 | |
| Foxi3 | |
| A synthetic peptide corresponding to internal region of mouse Foxi3. | |
| 100 μg | |
| Primary | |
| Mouse, Rat | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction