Learn More
Invitrogen™ GLYR1 Polyclonal Antibody

Description
The antibody detects endogenous levels of total GLYR1 protein.
Specifications
Specifications
| Antigen | GLYR1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1.4 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | GLYR1 |
| Gene Accession No. | Q49A26, Q5RKH0, Q922P9 |
| Gene Alias | 2810419J22Rik; 3930401k13rik; 3-hydroxyisobutyrate dehydrogenase-like protein; AW545332; BM045; Cytokine-like nuclear factor N-PAC; glyoxylate reductase 1 homolog; glyoxylate reductase 1 homolog (Arabidopsis); GLYR1; HIBDL; hNDF; NDF; NP60; Npac; N-pac; nuclear protein 60 kDa; nuclear protein 60kDa; nuclear protein NP60; Nuclear protein of 60 kDa; Nucleosome-destabilizing factor; Putative oxidoreductase GLYR1; RGD1309459 |
| Gene Symbols | GLYR1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.