Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GluR4 (extracellular) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA577424
Description
Product is shipped at room temperature as a lyophilized powder and should be stored at -20 °C upon receipt. Reconstitution: add 50 μL of deionized water.
Glutamate receptors mediate most excitatory neurotransmission in the brain and play an important role in neural plasticity, neural development and neurodegeneration. Ionotropic glutamate receptors are categorized into NMDA receptors and kainate/AMPA receptors, both of which contain glutamate-gated, cation-specific ion channels. Kainate/AMPA receptors are co-localized with NMDA receptors in many synapses and consist of seven structurally related subunits designated GluR-1 to -7. The kainate/AMPA receptors are primarily responsible for the fast excitatory neuro-transmission by glutamate, whereas the NMDA receptors are functionally characterized by a slow kinetic and a high permeability for Ca2+ ions.
Specifications
| GluR4 (extracellular) | |
| Polyclonal | |
| Unconjugated | |
| GRIA4 | |
| GRIA4, GLUA4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 14802, 2893, 29629 | |
| -20°C | |
| Lyophilized |
| Immunofluorescence, Immunohistochemistry, Western Blot | |
| 0.6 mg/mL | |
| PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
| P19493, P48058, Q9Z2W8 | |
| Gria4 | |
| Peptide (C)KLMDRWKKLDQRE, corresponding to amino acid residues 274-286 of rat AMPA Receptor 4 | |
| 50 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction