Learn More
                                    BYK Gardner GLOSS METER F/GP SEMI GLOSS
Supplier: BYK Gardner 4561
            Gloss meter for general purpose - semi gloss surfaces.Single angle 60°, especially for semi gloss surfacesFor any material: paint, plastic or even metalsMeasuring area 9 x 15 mm
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.