Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Trypsin-EDTA (0.25%), phenol red
GREENER_CHOICE

Catalog No. 25200114
Encompass_Preferred
Click to view available options
Quantity:
100 mL
500 mL
20 x 100 mL

Catalog No. 25-200-114

Supplier: Gibco™ 25200114

Only null left
Add to Cart
Edge
Add to Cart

Made from trypsin powder, irradiated mixture of proteases derived from porcine pancreas

Due to its digestive strength, trypsin is widely used for cell dissociation, routine cell culture passaging, and primary tissue dissociation. The trypsin concentration required for dissociation varies with cell type and experimental requirements.

  • Solutions tested for pH, osmolality, sterility, and performance
  • Raw materials are verified for e-beam irradiation and tested for endotoxin, PPV, PCV 1/2, mycoplasma, bacterial, fungal, and viral contamination, as well as multiple activity assays, ash analysis, and moisture analysis
  • Documented traceability available including lot traceability, animal origin certificates, lot analyses, irradiation certificates, viral inactivation summary, and supply chain transparency

Cell Culture, Mammalian Cell Culture

Order Info

Shipping Condition: Dry Ice

Compliance

Manufactured at a cGMP compliant facility registered with the FDA as a medical device manufacturer and certified to ISO 13485 standards.

TRUSTED_SUSTAINABILITY

Specifications

Chelators EDTA
Content And Storage Storage conditions: -5°C to -20°C
Shipping conditions: Wet ice
Shelf life: 24 months from date of manufacture
Cell Type Mammalian Cells
Classification Animal Origin
Concentration 1 X
Form Liquid
Product Type Cell Culture Dissociation Reagent
Sterility Sterile-filtered
Sterilization Method Sterile-filtered
With Additives Phenol Red, EDTA
Osmolality 270 - 320 mOsm/kg
pH 7.1 to 8
Product Line Gibco
Quantity 20 x 100 mL
Source Animal Origin
Tested For In vitro Bioassay
Show More Show Less

For Research Use or Further Manufacturing. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.