Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ TrypLE™ Express Enzyme (1X), phenol red
GREENER_CHOICE

Catalog No. 12605036
Encompass_Preferred
Click to view available options
Quantity:
100 mL
500 mL
5 L
20 x 100mL

Catalog No. 12-605-036

Supplier: Gibco™ 12605036

Only null left
Add to Cart
Add to Cart

Animal origin-free, recombinant enzyme

Used for dissociating a wide range of adherent mammalian cells, including CHO, HEK 293, A529, primary human keratinocytes, and embryonic stem cells. TrypLE™ Express cleaves peptide bonds on the C-terminal sides of lysine and arginine, and is a direct replacement for trypsin. Exceptional purity increases specificity and reduces damage to cells that can be caused by other enzymes present in some trypsin extracts.

  • Gentle On Cells
  • urity increases specificity due to the action of single enzyme, reducing damage caused by cleavage from enzymes present in some trypsin preparations and in other protease extracts
  • Room-Temperature Stable for 24 months
  • Ready to use
  • Can be directly substituted for trypsin in existing protocols
  • Dilution alone inactivates TrypLE Express, avoiding the need for trypsin inhibitors, such as FBS

Energy efficient, less hazardous, sustainable packaging

Cell Culture, Mammalian Cell Culture

Order Info

Shipping Condition: Room Temperature

Compliance

Manufactured in facilities compliant with cGMP manufacturing requirements, certified to the ISO 13485 standard, and registered with the FDA as medical device manufacturers.

TRUSTED_SUSTAINABILITY

Specifications

Chelators EDTA
Content And Storage Storage conditions: 15°C to 30°C, protect from light
Alternate storage conditions: 2°C to 8°C or -5°C to -20°C
Shipping conditions: Room temperature
Shelf life: 24 months from date of manufacture
Cell Type Mammalian Cells
Classification Animal Origin-free
Concentration 1 X
Culture Type Adherent Cell Culture
Form Liquid
Product Type Cell Culture Dissociation Reagent
Sterility Sterile-filtered
With Additives Phenol Red, EDTA
Green Features Energy efficient, Sustainable packaging
Osmolality Check and Record
Product Line Gibco
Quantity 20 x 100mL
Remarks Room Temperature Stable
Shipping Condition Room Temperature
Source Animal origin-free
Tested For In vitro Bioassay
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.