Learn More
Gibco™ MEM, NEAA, no glutamine

Description
Minimum Essential Medium (MEM) is one of the most commonly used of all cell culture media. MEM can be used with a variety of suspension and adherent mammalian cells, including HeLa, BHK-21, 293, HEP-2, HT-1080, MCF-7, fibroblasts, and primary rat astrocytes. We offer a variety of Gibco™ MEM modifications for a range of cell culture applications. Find the right formulation using the media selector tool.
This MEM is modified as follows:
With: NEAA, Phenol Red
Without: L-glutamine, HEPES
The complete formulation is available.
Gibco™ MEM, developed by Harry Eagle, was based on his earlier formulation of Basal Medium Eagle (BME). Many other modifications of MEM followed, including Glasgow's MEM, MEM α, DMEM, and Temin's Modification. MEM is available with Earle's salts for use in a CO2 incubator, or with Hanks' salts for use without CO2. This product is made with Earle's salts.
MEM contains no proteins, lipids, or growth factors. Therefore, MEM requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). MEM uses a sodium bicarbonate buffer system (2.2 g/L) and therefore requires a 5–10% CO2 environment to maintain physiological pH.
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Cell Line | HeLa, BHK-21, 293, HEP-2, HT-1080, MCF-7, and fibroblasts |
| Cell Type | Primary Rat Astrocytes |
| Classification | Animal Origin-free |
| Concentration | 1 X |
| Form | Liquid |
| Product Type | MEM (Minimum Essential Medium) |
| Serum Level | Standard Serum Supplementation |
| Sterility | Sterile-filtered |
| Sterilization Method | Sterile-filtered |
| With Additives | Low Glucose, Phenol Red, Non Essential Amino Acids (NEAA) |
| Show More |
Safety and Handling
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.