Learn More
SKC Inc GHOST WIPES 200/PK 2252414
Supplier: SKC Inc 2252414
SKC Ghost Wipes, For surface lead or surface dust sampling, Meets ASTM E1792 specifications and the new AIHA policy on sampling materials for lead in surface dust DOES NOT ship to Alaska, Arizona, California, Colorado, Hawaii, Idaho, Nevada, Oregon, Utah, Washington. Wyoming”
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.