Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Justrite™ Gas Cylinder Barricade Rack
Shop All Justrite Safety Group Products
Click to view available options
Holds:
1 Cylinder: 1 Wide by 1 Deep
10 Cylinders: 5 Wide by 2 Deep
12 Cylinders: 3 Wide by 4 Deep
12 Cylinders: 4 Wide by 3 Deep
15 Cylinders: 5 Wide by 3 Deep
16 Cylinders: 4 Wide by 4 Deep
2 Cylinders: 1 Wide by 2 Deep
2 Cylinders: 2 Wide by 1 Deep
20 Cylinders: 5 Wide by 4 Deep
3 Cylinders: 1 Wide by 3 Deep
3 Cylinders: 3 Wide by 1 Deep
4 Cylinders: 1 Wide by 4 Deep
4 Cylinders: 2 Wide by 2 Deep
4 Cylinders: 4 Wide by 1 Deep
5 Cylinders: 5 Wide by 1 Deep
6 Cylinders: 2 Wide by 3 Deep
6 Cylinders: 3 Wide by 2 Deep
8 Cylinders: 2 Wide by 4 Deep
8 Cylinders: 4 Wide by 2 Deep
9 Cylinders: 3 Wide by 3 Deep
Description
- Meets USGS high-hazard seismic zone requirements when properly mounted
- Recommended for indoor or outdoor bulk cylinder storage
- Surround cylinders with 2-in (51-mm) square steel structural tubing
- Continuously seam welded at all junction points
- Sealed with exterior grade powder-coat paint for long lasting strength, durability and dependability
- Tops ground smooth and capped with high-density polyethylene (HDPE) caps to prevent corrosion and injuries
- Features two levels of welded steel chains for each cylinder and clasps to connect the chains to the rack, ensuring a snug fit
- Pre-drilled 0.531-in (14-mm) diameter floor mounting holes included on lower tube structure
- For seismic installations, add Model 35350 S4-rated anchor bolts (not included)
Compliance
OSHA, CGA P1, NFPA55
Warranty and Services
1 YEAR
Specifications
Specifications
| Holds | 5 Cylinders: 5 Wide by 1 Deep |
| Material | Steel |
| Color | Black |
| For Use With (Equipment) | Gas cylinder |
| Height (Metric) | 762 mm |
| Length (Metric) | 1778 mm |
| Weight (English) | 115 lb. |
| Width (Metric) | 1778 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction