Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

gamma-Synuclein Antibody (SAIC-31A-10), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB275597 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB275597 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB275597 Supplier Novus Biologicals Supplier No. NBP320099MFV610
Only null left
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

gamma-Synuclein Monoclonal antibody specifically detects gamma-Synuclein in Human samples. It is validated for Flow Cytometry

Specifications

Antigen gamma-Synuclein
Applications Flow Cytometry
Classification Monoclonal
Clone SAIC-31A-10
Conjugate mFluor Violet 610 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias BCSG1PERSYN, Breast cancer-specific gene 1 protein, gamma-synuclein, persyn, SRPRSN, synoretin, synuclein, gamma (breast cancer-specific protein 1)
Host Species Rabbit
Immunogen The KLH-conjugated peptide YLAPSGPSGTLK, derived from Gamma-synuclein. Gamma-synulein plays a role in neurofilament network integrity.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Cancer, Cellular Markers, Core ESC Like Genes, Mitotic Regulators, Neuroscience, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 6623
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.