Learn More
Specifications
Specifications
| Packaging | Natural Poly Bottle |
| CAS Min % | 98.0, 0.11 |
| CAS Max % | 100.0, 0.11 |
| Physical Form | Liquid |
| Color | Colorless |
| Melting Point | 0.0°C |
| Boiling Point | 100°C |
| For Use With (Equipment) | GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation |
| Solubility | Miscible with water |
| Solution Type | Calibration Standard |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
