Learn More
Bacharach™ Fyrite™ CO2 Gas Analyzers
Click to view available options
For Use With (Application):
Measuring and analyzing carbon dioxide
Description
Includes
- Sampling assembly
- Carrying case
Fast, accurate and easy-to-use instrument for measuring and analyzing carbon dioxide
- The number of tests possible with one fluid charge depends on the concentration of samples being tested; at midpoint scale reading the CO2 fluid is good for approximately 300 gas samples and the O2 fluid for 100 tests
- Indicators may be exposed to ambient temperatures from -30° to +150°F, and gases up to 850°F may be tested with standard aspirator sampling equipment (special sampling equipment for higher gas temperatures or dry gases is available)
- The need to replace fluid can be easily determined with a simple test, and replacement is an easy procedure
- For temperatures above 1400°F, a ceramic sampling tube is available
- Readings are unaffected by the presence of most background gases in the sample
- Produced in three scale ranges
- Accuracy is within ±1/2% CO2
Order Info
Kits shipped without fluid
Specifications
Specifications
| Type | CO2 Single Kit |
| For Use With (Application) | Measuring and analyzing carbon dioxide |
| Includes | Analyzer, sampling assembly and carrying case. (Indicator sold separately.) |