Learn More
AG Scientific Inc Formalin 500 ML
Supplier: AG Scientific Inc F2467500ML
Formalin 10% Neutral Buffered Solution is a widely used tissue fixative for common histology applications. Ready to use formula does not require dilution.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.