Learn More
World Precision Instrument Micro Mosquito Hemostatic Forceps, 12.5 cm, Curved
SDP
Supplier:  World Precision Instrument 500452
Encompass
Micro Mosquito Forceps, also known as microsurgical mosquito forceps or micro hemostatic forceps, are designed for extremely delicate and precise surgical procedures. Micro Mosquito Forceps are significantly smaller in size compared to regular mosquito forceps. These are 12.5 cm (5") long with fine tips. Looking at the end of the tip of one blade of these forceps, the dimensions are 0.4 mm thick and 0.8 mm wide.
Catalog No. 50-822-518
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.